Learn More
MSC PROPPER Flight Suit, Nomex, Size 40 Short, Green, 1 ea.
Catalog No. 5000768700
Encompass
This flight suit is constructed from Nomex material, offering flame-resistant and flame-retardant properties for enhanced protection. Designed for military specifications, it features a zipper closure and multiple pockets for functionality.
  • Adjustable waist belt with hook and loop closure
  • Bi-swing back for increased mobility
  • Knife pocket on left leg inseam
  • Sewn to military specification FNS/PD 96-17 (MIL-C-83141A)
  • Open cuff and ankle style
  • Flat felled seams
Catalog No. 50-007-68700 Supplier MSC Supplier No. 54612890
This flight suit is constructed from Nomex material, offering flame-resistant and flame-retardant properties for enhanced protection. Designed for military specifications, it features a zipper closure and multiple pockets for functionality.
  • Adjustable waist belt with hook and loop closure
  • Bi-swing back for increased mobility
  • Knife pocket on left leg inseam
  • Sewn to military specification FNS/PD 96-17 (MIL-C-83141A)
  • Open cuff and ankle style
  • Flat felled seams