Learn More
Enzo Life Sciences Propranolol (5 g). CAS: 318-98-9
SDP
Supplier:  Enzo Life Sciences BMLST4050005
Encompass_Preferred
Propranolol blocks PLD derived DAG formation by inhibiting phosphatidate phosphohydrolase (IC50=70 µM). It is a useful tool to discriminate between PLC or PLD mediated DAG production. It is also a β-adrenergic receptor blocking agent. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-2180