Learn More
Medchemexpress LLC Propranolol-d7 hydrochloride | 1613439-56-7 | MFCD11983489 | 98.5% | 10 MG
SDP
Catalog No. 5000298074
Encompass_Preferred
Propranolol-d7 hydrochloride is a deuterium-labeled analog of propranolol supplied as a solid hydrochloride salt. It is intended for use as a labeled reference standard and internal standard in LC-MS/MS and bioanalytical assays for quantitation and pharmacokinetic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Hydrochloride salt, solid form for stable storage.
  • High isotopic labeling and ≥98.5% chemical purity.
  • Suitable for LC-MS/MS quantitation and pharmacokinetic studies.
  • Provided in small reference quantities for analytical workflows.
Catalog No. 50-002-98074 Supplier Medchemexpress LLC Supplier No. HYB0573S10MG
Propranolol-d7 hydrochloride is a deuterium-labeled analog of propranolol supplied as a solid hydrochloride salt. It is intended for use as a labeled reference standard and internal standard in LC-MS/MS and bioanalytical assays for quantitation and pharmacokinetic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Hydrochloride salt, solid form for stable storage.
  • High isotopic labeling and ≥98.5% chemical purity.
  • Suitable for LC-MS/MS quantitation and pharmacokinetic studies.
  • Provided in small reference quantities for analytical workflows.