Learn More
Medchemexpress LLC Propranolol glycol | 36112-95-5 | MFCD00673941 | 99.3% | 218.25 g/mol | C13H14O3 | 25 MG
SDP
Catalog No. 5000212852
Encompass_Preferred
Propranolol glycol is an endogenous metabolite of propranolol used in research to study drug metabolism and anticonvulsant activity. It is supplied as a purified chemical standard for analytical, pharmacological, and in vivo studies, with associated safety and quality documentation for proper handling.
  • Metabolite with reported anticonvulsant activity.
  • High purity suitable for analytical and pharmacological studies.
  • Supplied as a stable solid standard for in vitro and in vivo research.
  • Characterized by CAS 36112-95-5 and molecular weight 218.25 g/mol.
  • Includes SDS and certificate of analysis for quality verification.
  • Appropriate for drug metabolism and seizure-model investigations.
Catalog No. 50-002-12852 Supplier Medchemexpress LLC Supplier No. HY10124025MG
Propranolol glycol is an endogenous metabolite of propranolol used in research to study drug metabolism and anticonvulsant activity. It is supplied as a purified chemical standard for analytical, pharmacological, and in vivo studies, with associated safety and quality documentation for proper handling.
  • Metabolite with reported anticonvulsant activity.
  • High purity suitable for analytical and pharmacological studies.
  • Supplied as a stable solid standard for in vitro and in vivo research.
  • Characterized by CAS 36112-95-5 and molecular weight 218.25 g/mol.
  • Includes SDS and certificate of analysis for quality verification.
  • Appropriate for drug metabolism and seizure-model investigations.