Learn More
Medchemexpress LLC Propranolol glycol | 36112-95-5 | MFCD00673941 | 99.3% | 218.25 g/mol | C13H14O3 | 5 MG
SDP
Catalog No. 5000211749
Encompass_Preferred
Propranolol glycol is an endogenous metabolite of propranolol that exhibits rapid anticonvulsant activity and is used as a research-grade analytical standard in studies of drug metabolism and pharmacology.
  • Endogenous metabolite of propranolol.
  • Instantaneous anticonvulsant activity reported in literature.
  • High purity suitable for analytical and research use.
  • Available in small-quantity formats for use as standards.
  • Stable when stored under recommended conditions for reliable results.
Catalog No. 50-002-11749 Supplier Medchemexpress LLC Supplier No. HY1012405MG
Propranolol glycol is an endogenous metabolite of propranolol that exhibits rapid anticonvulsant activity and is used as a research-grade analytical standard in studies of drug metabolism and pharmacology.
  • Endogenous metabolite of propranolol.
  • Instantaneous anticonvulsant activity reported in literature.
  • High purity suitable for analytical and research use.
  • Available in small-quantity formats for use as standards.
  • Stable when stored under recommended conditions for reliable results.