Learn More
Eurofins Discoverx Propranolol Hydrochloride, 10 mg. (92-1247)
Supplier:  Eurofins Discoverx 921247
Encompass
Propranolol is a sympatholytic beta-adrenergic antagonist, widely used for treatment of tremors, angina, hypertension, heart rhythm disorders, migraines, thyrotoxicosis, anxiety, and other circulatory diseases. It was the first successful beta-blocker developed. Acts as an antagonist for ADRAB1 and ADRAB2. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9768