Learn More
Medchemexpress LLC HY-B0573 1g , Propranolol (hydrochloride) CAS:318-98-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB05731G
Encompass_Preferred
HY-B0573 1g Propranolol (hydrochloride) CAS: 318-98-9 Propranolol hydrochloride is a nonselective β-adrenergic receptor (βAR) antagonist, has high affinity for the β1AR and β2AR with Ki values of 1.8 nM and 0.8 nM, respectively. Propranolol hydrochloride inhibits [3H]-DHA binding to rat brain membrane preparation with an IC50 of 12 nM. Propranolol hydrochloride is used for study of hypertension, pheochromocytoma, myocardial infarction, cardiac arrhythmias, angina pectoris, and hypertrophic cardiomyopathy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9280