Learn More
Apexbio Technology LLC Propranolol HCl 318-98-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089806
Encompass_Preferred
Propranolol HCl (CAS 318-98-9) is a small-molecule antagonist targeting 1- and 2-adrenergic receptors It is designed to competitively inhibit beta-adrenergic receptors thereby modulating sympathetic nervous system signaling Propranolol HCl exerts its biological activity primarily through competitive blockade of -adrenergic receptors preventing the binding of endogenous catecholamines In in vitro studies propranolol HCl demonstrates inhibitory activity with IC50 values typically within the range of 10 50 nM for -adrenoceptor binding assays Based on these pharmacological properties propranolol HCl holds research potential in cardiovascular regulatory studies neurotransmission models stress response experiments and tumor-related angiogenesis assays
Catalog No. 50-000-89806 Supplier Apexbio Technology LLC Supplier No. B13465.1
Propranolol HCl (CAS 318-98-9) is a small-molecule antagonist targeting 1- and 2-adrenergic receptors It is designed to competitively inhibit beta-adrenergic receptors thereby modulating sympathetic nervous system signaling Propranolol HCl exerts its biological activity primarily through competitive blockade of -adrenergic receptors preventing the binding of endogenous catecholamines In in vitro studies propranolol HCl demonstrates inhibitory activity with IC50 values typically within the range of 10 50 nM for -adrenoceptor binding assays Based on these pharmacological properties propranolol HCl holds research potential in cardiovascular regulatory studies neurotransmission models stress response experiments and tumor-related angiogenesis assays