Learn More
AdipoGen Propranolol HCl
SDP
Catalog No. 502560190
Encompass_Preferred
Chemical. CAS 318-98-9. Formula C16H21NO2 . HCl. Molecular Weight 295.8. ±-Propranolol hydrochloride is a beta-adrenergic receptor beta-AR antagonist Kds = 6.91, 0.832, and 117.49 nM for human beta1-, beta2- and beta3-ARs, respectively. It also acts as a non-specific 5-HT1/5-HT2 serotonin receptor antagonist, reducing tranylcypromine/L-tryptophan-induced hyperactivity in rats. ±-Propranolol hydrochloride is useful as an antihypertensive drug, cardiac depressant and also in the treatment of angina pectoris. It decreases the effect of stress and exercise on heart by reducing the rate of contraction and conduction of impulse. It is known to competitively block the action of catecholamines.
Catalog No. 50-256-0190 Supplier AdipoGen Supplier No. CDXP0483G100
Chemical. CAS 318-98-9. Formula C16H21NO2 . HCl. Molecular Weight 295.8. ±-Propranolol hydrochloride is a beta-adrenergic receptor beta-AR antagonist Kds = 6.91, 0.832, and 117.49 nM for human beta1-, beta2- and beta3-ARs, respectively. It also acts as a non-specific 5-HT1/5-HT2 serotonin receptor antagonist, reducing tranylcypromine/L-tryptophan-induced hyperactivity in rats. ±-Propranolol hydrochloride is useful as an antihypertensive drug, cardiac depressant and also in the treatment of angina pectoris. It decreases the effect of stress and exercise on heart by reducing the rate of contraction and conduction of impulse. It is known to competitively block the action of catecholamines.