Learn More
Selleck Chemical LLC Propranolol HCl S4076-1g
SDP
Catalog No. 503109495
Encompass_Preferred
Propranolol HCl (AY-64043 ICI-45520 NCS-91523) is a competitive non-selective beta-adrenergic receptors inhibitor
Catalog No. 50-310-9495 Supplier Selleck Chemical LLC Supplier No. S40761G
Propranolol HCl (AY-64043 ICI-45520 NCS-91523) is a competitive non-selective beta-adrenergic receptors inhibitor