Learn More
Medchemexpress LLC Propranolol (hydrochloride) | 318-98-9 | 99.9% | 1 G
SDP
Catalog No. 5000407914
Encompass_Preferred
Propranolol is a nonselective and BBB-permeable β-adrenergic receptor (βAR) antagonist with high affinity for β1AR and β2AR. It inhibits [3H]-DHA binding to rat brain membrane preparation. This product is utilized in studies concerning hypertension, pheochromocytoma, myocardial infarction, cardiac arrhythmias, angina pectoris, and hypertrophic cardiomyopathy.
  • Nonselective and BBB-permeable β-adrenergic receptor antagonist
  • Exhibits high affinity for β1AR and β2AR
  • Inhibits [3H]-DHA binding to rat brain membrane preparation
  • Increases total ERK1/2 levels in HemSCs
  • Decreases cell proliferation in HemSCs
  • Induces HemSC apoptosis
  • Reduces vessel diameter in IH Matrigel implant model
  • Can be used to induce psoriasis models
Catalog No. 50-004-07914 Supplier Medchemexpress LLC Supplier No. HYB0573B1G
Propranolol is a nonselective and BBB-permeable β-adrenergic receptor (βAR) antagonist with high affinity for β1AR and β2AR. It inhibits [3H]-DHA binding to rat brain membrane preparation. This product is utilized in studies concerning hypertension, pheochromocytoma, myocardial infarction, cardiac arrhythmias, angina pectoris, and hypertrophic cardiomyopathy.
  • Nonselective and BBB-permeable β-adrenergic receptor antagonist
  • Exhibits high affinity for β1AR and β2AR
  • Inhibits [3H]-DHA binding to rat brain membrane preparation
  • Increases total ERK1/2 levels in HemSCs
  • Decreases cell proliferation in HemSCs
  • Induces HemSC apoptosis
  • Reduces vessel diameter in IH Matrigel implant model
  • Can be used to induce psoriasis models