Learn More
Biotium Propidium Iodide, 1 mg/mL solution in water
Catalog No. NC0221356
Encompass_Preferred
Propidium iodide (PI) is a cell impermeable nucleic acid intercalating dye. Because PI is excluded from viable cells it can be used in apoptotic studies to distinguish necrotic cells or cells in the late stages of apoptosis from viable cells or cells in early apoptosis where cell membranes are still intact. PI is also utilized as a counterstain in multicolor fluorescent imaging. After entering cells and binding to DNA and RNA, the fluorescence of PI is enhanced 20- to 30-fold. PI can be used in flow cytometry and fluorescence microscopy. 1 mg/mL solution in water Orange / red liquid Store at 4°C and protect from light &lamba;Ex/&lamba;Em (DNA-bound) = 535/617 nm &lamba;Ex/&lamba;Em (no DNA) = 493/636 nm C27H34I2N4 MW: 668.4 [25535-16-4]
Catalog No. NC0221356 Supplier Biotium Supplier No. 40017
Propidium iodide (PI) is a cell impermeable nucleic acid intercalating dye. Because PI is excluded from viable cells it can be used in apoptotic studies to distinguish necrotic cells or cells in the late stages of apoptosis from viable cells or cells in early apoptosis where cell membranes are still intact. PI is also utilized as a counterstain in multicolor fluorescent imaging. After entering cells and binding to DNA and RNA, the fluorescence of PI is enhanced 20- to 30-fold. PI can be used in flow cytometry and fluorescence microscopy. 1 mg/mL solution in water Orange / red liquid Store at 4°C and protect from light &lamba;Ex/&lamba;Em (DNA-bound) = 535/617 nm &lamba;Ex/&lamba;Em (no DNA) = 493/636 nm C27H34I2N4 MW: 668.4 [25535-16-4]