Learn More
Hach Company GLI Proprietary Gel, 2 Grams
Catalog No. 501997426 Shop All Hach Company Products
Encompass
Chem.Proprietary Gel Powder for pHD sc and pHD (2 grams; to gel standard cell solution for high temperature applications).
Catalog No. 50-199-7426 Supplier Hach Company Supplier No. 25M8A1002101
Chem.Proprietary Gel Powder for pHD sc and pHD (2 grams; to gel standard cell solution for high temperature applications).