Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proprietary Solvent III-1, Anhydrous, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003890 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-890 4 L Narrow Mouth PVC-coated Amber Bottle
02-003-889 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-003-890 Supplier Avantor Supplier No. 928733
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Specifications

Chemical Name or Material Proprietary Solvent III-1
Name Note Anhydrous
Quantity 4 L
Flash Point 13.00°C
Packaging Narrow Mouth PVC-coated Amber Bottle
Specific Gravity 1L = 0.79kg
Grade BAKER ANALYZED™ Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.