Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019344 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9344 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9344 Supplier MilliporeSigma Supelco Supplier No. 408585ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 842°F
CAS 109-60-4
Vapor Pressure 25 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.888 g/mL (at 25°C)
Flash Point 14°C
Physical Form Neat
Linear Formula CH3 COOCH2 CH2 CH3
MDL Number MFCD00009372
Melting Point -95°C
Boiling Point 102°C
Solubility Soluble in water
Type Analytical Standard
Quantity 5 mL
Formula Weight 102.13
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.