Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Propyl benzoate, 99%

Catalog No. AAH6679330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6679330 250 g
AAH6679318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6679330 Supplier Thermo Scientific™ Supplier No. H6679330
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2315-68-6 | C10H12O2 | 164.20 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-68-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009370
InChI Key UDEWPOVQBGFNGE-UHFFFAOYSA-N
IUPAC Name propyl benzoate
SMILES CCCOC(=O)C1=CC=CC=C1

Specifications

Density 1.026
Boiling Point 231°
Flash Point 98°(208°F)
Linear Formula C{6}H{5}CO{2}CH{2}CH{2}CH{3}
Quantity 250 g
Formula Weight 164.2
Physical Form Liquid
ShelfLife 5 Years
EINECSNumber 219-020-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.