Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Benzoate 99.0+%, TCI America™
SDP

Catalog No. B007625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007625ML 25 mL
B0076500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B007625ML Supplier TCI America Supplier No. B007625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2315-68-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009370
InChI Key UDEWPOVQBGFNGE-UHFFFAOYSA-N
Synonym n-propyl benzoate, benzoic acid, propyl ester, benzoic acid n-propyl ester, propyl benzenecarboxylate, benzoic acid propyl ester, unii-vwk210b7ws, benzoate de propyle, vwk210b7ws, dsstox_cid_24878, dsstox_rid_80552
PubChem CID 16846
IUPAC Name propyl benzoate
SMILES CCCOC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Benzoate
EINECSNumber (3)-1356
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.