Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl dodecanoate | 3681-78-5 | 242.40 | 1 ML
SDP

Catalog No. 5000449560
Encompass_Preferred

Propyl dodecanoate, also known as Propyl laurate, is an ester product. Intended for research use only, this liquid appears colorless to light yellow, offering high purity for various laboratory applications.

  • Purity: 99.8%
  • Molecular weight: 242.40
  • CAS number: 3681-78-5
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage stability: 3 years at room temperature
  • Storage in solvent: 2 years at -80°C or 1 year at -20°C
  • Solubility in DMSO: 100 mg/mL
  • In vivo solubility: ≥ 5 mg/mL in specific protocols

Catalog No. 50-004-49560 Supplier Medchemexpress LLC Supplier No. HYW00227010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl dodecanoate, also known as Propyl laurate, is an ester product. Intended for research use only, this liquid appears colorless to light yellow, offering high purity for various laboratory applications.

  • Purity: 99.8%
  • Molecular weight: 242.40
  • CAS number: 3681-78-5
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage stability: 3 years at room temperature
  • Storage in solvent: 2 years at -80°C or 1 year at -20°C
  • Solubility in DMSO: 100 mg/mL
  • In vivo solubility: ≥ 5 mg/mL in specific protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.