Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl dodecanoate | 3681-78-5 | 242.40 | 100 G
SDP

Catalog No. 5000449045
Encompass_Preferred

Propyl dodecanoate, also known as Propyl laurate, is an ester product provided by MedChemExpress. This compound features a molecular weight of 242.40 and a high purity of 99.8%. It is supplied as a colorless to light yellow liquid and is intended strictly for research use.

  • Ester product
  • Purity of 99.8%
  • Molecular weight of 242.40
  • Molecular formula C15H30O2
  • Liquid appearance, colorless to light yellow
  • Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Catalog No. 50-004-49045 Supplier Medchemexpress LLC Supplier No. HYW002270100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl dodecanoate, also known as Propyl laurate, is an ester product provided by MedChemExpress. This compound features a molecular weight of 242.40 and a high purity of 99.8%. It is supplied as a colorless to light yellow liquid and is intended strictly for research use.

  • Ester product
  • Purity of 99.8%
  • Molecular weight of 242.40
  • Molecular formula C15H30O2
  • Liquid appearance, colorless to light yellow
  • Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.