Learn More
Medchemexpress LLC Propyl dodecanoate | 3681-78-5 | 242.40 | 25 G
SDP
Catalog No. 5000448189
Encompass_Preferred
Propyl dodecanoate, also known as Propyl laurate, is characterized as an ester product. This compound is a colorless to light yellow liquid with a molecular weight of 242.40 and a chemical formula of C15H30O2. It is intended for research use only and is soluble in DMSO at concentrations up to 100 mg/mL, requiring sonication for dissolution.
Features and benefits:
  • It is an ester product.
  • It appears as a colorless to light yellow liquid.
  • It is soluble in DMSO.
  • It is suitable for research applications.
Catalog No. 50-004-48189 Supplier Medchemexpress LLC Supplier No. 25GHYW002270
Propyl dodecanoate, also known as Propyl laurate, is characterized as an ester product. This compound is a colorless to light yellow liquid with a molecular weight of 242.40 and a chemical formula of C15H30O2. It is intended for research use only and is soluble in DMSO at concentrations up to 100 mg/mL, requiring sonication for dissolution.
Features and benefits:
  • It is an ester product.
  • It appears as a colorless to light yellow liquid.
  • It is soluble in DMSO.
  • It is suitable for research applications.