Learn More
Cayman Chemical Propyl Gallate, Purity: ≥98%, CAS: 121-79-9, 500 g
SDP
Catalog No. 502964290
Encompass_Preferred
A phenolic antioxidant with antimicrobial activity; hepatoprotective in vitro and in vivo, preventing CCl4 induced lipoperoxidation and reduction in polysomes in rat liver; increases expression of HIF-1α, EPO, and VEGF mRNA levels and the number of normal neurons in rat brains after forebrain ischemia; commonly added to foods to prevent autoxidation and microbial growth
Catalog No. 50-296-4290 Supplier Cayman Chemical Supplier No. 22293500 G
A phenolic antioxidant with antimicrobial activity; hepatoprotective in vitro and in vivo, preventing CCl4 induced lipoperoxidation and reduction in polysomes in rat liver; increases expression of HIF-1α, EPO, and VEGF mRNA levels and the number of normal neurons in rat brains after forebrain ischemia; commonly added to foods to prevent autoxidation and microbial growth