Learn More
Medchemexpress LLC Propyl gallate | 121-79-9 | 212.20 | 5 G
SDP
Catalog No. 5000390098
Encompass_Preferred
Propyl gallate is a common food antioxidant that can inhibit the production of acrolein, glyoxal, and methylglyoxal. It exhibits anti-inflammatory, antitumor, and cardioprotective activities.
  • Common food antioxidant.
  • Can inhibit the production of acrolein, glyoxal, and methylglyoxal.
  • Exhibits anti-inflammatory activities.
  • Exhibits antitumor activities.
  • Exhibits cardioprotective activities.
Catalog No. 50-003-90098 Supplier Medchemexpress LLC Supplier No. HYN05245G
Propyl gallate is a common food antioxidant that can inhibit the production of acrolein, glyoxal, and methylglyoxal. It exhibits anti-inflammatory, antitumor, and cardioprotective activities.
  • Common food antioxidant.
  • Can inhibit the production of acrolein, glyoxal, and methylglyoxal.
  • Exhibits anti-inflammatory activities.
  • Exhibits antitumor activities.
  • Exhibits cardioprotective activities.