Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl gallate (Standard) | 121-79-9 | 212.20 | 50 MG
SDP

Catalog No. 5000389356
Encompass_Preferred

Propyl gallate (Standard) is the analytical standard of Propyl gallate. It is intended for research and analytical applications. Propyl gallate is a common food antioxidant that can inhibit the production of acrolein, glyoxal, and methylglyoxal. It also exhibits anti-inflammatory, antitumor, and cardioprotective activities.

  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Purity: 99.98%
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-89356 Supplier Medchemexpress LLC Supplier No. HYN0524R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl gallate (Standard) is the analytical standard of Propyl gallate. It is intended for research and analytical applications. Propyl gallate is a common food antioxidant that can inhibit the production of acrolein, glyoxal, and methylglyoxal. It also exhibits anti-inflammatory, antitumor, and cardioprotective activities.

  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Purity: 99.98%
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.