Learn More
Medchemexpress LLC Propyl gallate (standard) | 121-79-9 | MFCD00002196 | 212.20 | 100 MG
SDP
Catalog No. 5000390016
Encompass_Preferred
Propyl gallate (Standard) is the analytical standard of Propyl gallate, intended for research and analytical applications. It functions as a common food antioxidant and can inhibit the production of acrolein, glyoxal, and methylglyoxal. Additionally, it exhibits anti-inflammatory, antitumor, and cardioprotective activities.
  • Analytical standard for Propyl gallate
  • Intended for research and analytical applications
  • Common food antioxidant
  • Inhibits production of acrolein, glyoxal, and methylglyoxal
  • Exhibits anti-inflammatory activities
  • Exhibits antitumor activities
  • Exhibits cardioprotective activities
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-90016 Supplier Medchemexpress LLC Supplier No. HYN0524R100MG
Propyl gallate (Standard) is the analytical standard of Propyl gallate, intended for research and analytical applications. It functions as a common food antioxidant and can inhibit the production of acrolein, glyoxal, and methylglyoxal. Additionally, it exhibits anti-inflammatory, antitumor, and cardioprotective activities.
  • Analytical standard for Propyl gallate
  • Intended for research and analytical applications
  • Common food antioxidant
  • Inhibits production of acrolein, glyoxal, and methylglyoxal
  • Exhibits anti-inflammatory activities
  • Exhibits antitumor activities
  • Exhibits cardioprotective activities
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS