Learn More
Medchemexpress LLC Propyl hexanoate | 626-77-7 | MFCD00053803 | 99.90% | 158.24 | 50 G
SDP
Catalog No. 5000454706
Encompass_Preferred
Propyl hexanoate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Available as a liquid.
  • Appears colorless to off-white.
  • Shipped at room temperature in continental US.
  • Can be stored at room temperature for three years.
  • Stable in solvent at -80°C for two years or -20°C for one year.
Catalog No. 50-004-54706 Supplier Medchemexpress LLC Supplier No. HYW01361650G
Propyl hexanoate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Available as a liquid.
  • Appears colorless to off-white.
  • Shipped at room temperature in continental US.
  • Can be stored at room temperature for three years.
  • Stable in solvent at -80°C for two years or -20°C for one year.