Learn More
Medchemexpress LLC Propyl n-octanoate 50g
SDP
Catalog No. 5000206081
Encompass_Preferred
Propyl n-octanoate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry Furthermore it can also be used as a solvent or intermediate in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including perfumes lotions and hair care products
Catalog No. 50-002-06081 Supplier Medchemexpress LLC Supplier No. HYW12753550G
Propyl n-octanoate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry Furthermore it can also be used as a solvent or intermediate in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including perfumes lotions and hair care products