Learn More
Medchemexpress LLC HY-100689 10mg Medchemexpress, Propyl pyrazole triol CAS:263717-53-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10068910MG
Encompass_Preferred
Medchemexpress, HY-100689 10mg Propyl pyrazole triol CAS:263717-53-9 Propyl pyrazole triol (PPT) is an estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7769