Learn More
Selleck Chemical LLC Propyl pyrazole triol-25MG
SDP
Catalog No. 503137935
Encompass_Preferred
Propyl pyrazole triol is a selective estrogen receptor alpha (ER ) agonist
Catalog No. 50-313-7935 Supplier Selleck Chemical LLC Supplier No. E094025MG
Propyl pyrazole triol is a selective estrogen receptor alpha (ER ) agonist