Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl pyrazole triol | 263717-53-9 | 386.44 g/mol | C24H22N2O3 | 50 MG
SDP

Catalog No. 5000213547
Encompass_Preferred

Propyl pyrazole triol (PPT) is a synthetic, nonsteroidal selective estrogen receptor alpha (ERα) agonist used as a research reagent to probe ERα-mediated signaling in vitro and in vivo. It is supplied as a powder and includes manufacturer guidance for storage and formulation.

  • Selective estrogen receptor alpha (ERα) agonist with reported high ERα selectivity.
  • Intended for research use in vitro and in vivo studies.
  • Supplied as a powder, suitable for formulation into standard dosing vehicles.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Soluble in DMSO (≥100 mg/mL) and in recommended in vivo formulation vehicles.
  • Molecular weight 386.44 g/mol; CAS number 263717-53-9.

Catalog No. 50-002-13547 Supplier Medchemexpress LLC Supplier No. HY10068950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl pyrazole triol (PPT) is a synthetic, nonsteroidal selective estrogen receptor alpha (ERα) agonist used as a research reagent to probe ERα-mediated signaling in vitro and in vivo. It is supplied as a powder and includes manufacturer guidance for storage and formulation.

  • Selective estrogen receptor alpha (ERα) agonist with reported high ERα selectivity.
  • Intended for research use in vitro and in vivo studies.
  • Supplied as a powder, suitable for formulation into standard dosing vehicles.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Soluble in DMSO (≥100 mg/mL) and in recommended in vivo formulation vehicles.
  • Molecular weight 386.44 g/mol; CAS number 263717-53-9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.