Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl pyrazole triol | 263717-53-9 | 99.1% | 386.44 | C24H22N2O3 | 1 ML
SDP

Catalog No. 5000213546
Encompass_Preferred

Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist used in research to probe ERα-mediated signaling pathways. It is offered as solids in multiple sizes and as a ready-to-use 10 mM solution in DMSO (1 mL) to simplify in vitro assay setup. Documented purity, solubility, and storage data support reproducible experimental use.

  • Selective ERα agonist for receptor binding and functional studies.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexibility.
  • Reported purity 99.11% to support assay reliability.
  • Soluble in DMSO at ≥100 mg/mL, facilitating preparation of working solutions.
  • Storage recommendations provided for powder and solution to maintain stability.

Catalog No. 50-002-13546 Supplier Medchemexpress LLC Supplier No. HY10068910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist used in research to probe ERα-mediated signaling pathways. It is offered as solids in multiple sizes and as a ready-to-use 10 mM solution in DMSO (1 mL) to simplify in vitro assay setup. Documented purity, solubility, and storage data support reproducible experimental use.

  • Selective ERα agonist for receptor binding and functional studies.
  • Available as 10 mM solution in DMSO and multiple solid sizes for flexibility.
  • Reported purity 99.11% to support assay reliability.
  • Soluble in DMSO at ≥100 mg/mL, facilitating preparation of working solutions.
  • Storage recommendations provided for powder and solution to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.