Learn More
Medchemexpress LLC Propyl pyrazole triol | 263717-53-9 | 99.1% | 386.44 | C24H22N2O3
SDP
Catalog No. 5000214035
Encompass_Preferred
Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist supplied for research and analytical applications. Provided as a powder, it has confirmed identity (C24H22N2O3, CAS 263717-53-9, MW 386.44) and is intended for receptor binding and functional assays.
Catalog No. 50-002-14035 Supplier Medchemexpress LLC Supplier No. HY100689100MG
Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist supplied for research and analytical applications. Provided as a powder, it has confirmed identity (C24H22N2O3, CAS 263717-53-9, MW 386.44) and is intended for receptor binding and functional assays.