Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propyl pyrazole triol | 263717-53-9 | MFCD03453628 | 99.1% | 386.44 g·mol⁻¹ | C24H22N2O3 | 25 MG
SDP

Catalog No. 5000214175
Encompass_Preferred

Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist used in biochemical and cellular research to probe ERα-mediated signaling. It is a small-molecule phenolic compound with high selectivity for ERα over ERβ and is supplied as a solid for laboratory use. For research and analytical applications only; not for human use.

  • Selective ERα agonist for receptor-specific studies.
  • High ERα versus ERβ selectivity supported by binding data.
  • CAS number 263717-53-9 for unambiguous identification.
  • Molecular formula C24H22N2O3 and molecular weight 386.44 g·mol⁻¹.
  • Supplied as a solid powder suitable for solution preparation.
  • Intended for research and analytical applications only.

Catalog No. 50-002-14175 Supplier Medchemexpress LLC Supplier No. HY10068925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist used in biochemical and cellular research to probe ERα-mediated signaling. It is a small-molecule phenolic compound with high selectivity for ERα over ERβ and is supplied as a solid for laboratory use. For research and analytical applications only; not for human use.

  • Selective ERα agonist for receptor-specific studies.
  • High ERα versus ERβ selectivity supported by binding data.
  • CAS number 263717-53-9 for unambiguous identification.
  • Molecular formula C24H22N2O3 and molecular weight 386.44 g·mol⁻¹.
  • Supplied as a solid powder suitable for solution preparation.
  • Intended for research and analytical applications only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.