Learn More
Selleck Chemical LLC Propyl pyrazole triol-5MG
SDP
Catalog No. 503138856
Encompass_Preferred
Propyl pyrazole triol is a selective estrogen receptor alpha (ER ) agonist
Catalog No. 50-313-8856 Supplier Selleck Chemical LLC Supplier No. E09405MG
Propyl pyrazole triol is a selective estrogen receptor alpha (ER ) agonist