Learn More
Medchemexpress LLC HY-100689 5mg Medchemexpress, Propyl pyrazole triol CAS:263717-53-9 Purity:>98%
SDP
Catalog No. 501931917
Encompass_Preferred
Medchemexpress, HY-100689 5mg Propyl pyrazole triol CAS:263717-53-9 Propyl pyrazole triol (PPT) is an estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1917 Supplier Medchemexpress LLC Supplier No. HY1006895MG
Medchemexpress, HY-100689 5mg Propyl pyrazole triol CAS:263717-53-9 Propyl pyrazole triol (PPT) is an estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.