Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylbenzene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019749 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9749 5 mL
11-101-9015 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9749 Supplier MilliporeSigma Supelco Supplier No. 821185ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propylbenzene belongs to the class of aromatic hydrocarbons, commonly used as a solvent in organic synthesis and precursor to synthesize intermediates such as 4-phenylbutyraldehyde and ethyl phenyl ketone.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 103-65-1
Ignition Point 842°F
Vapor Pressure 2 mmHg (20°C)
Vapor Density 4.14 (vs. air)
Density 0.862 g/mL (at 25°C)
Flash Point 42°C
Physical Form Neat
Linear Formula C6H5 CH2 CH2 CH3
MDL Number MFCD00009377
Melting Point -99°C
Boiling Point 159°C
Synonym 1-Phenylpropane
Type Analytical Standard
Quantity 5 mL
Formula Weight 120.19
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.