Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propylboronic acid | 17745-45-8 | 107360 | 87.91 | 100 G
SDP

Catalog No. 5000460408
Encompass_Preferred

Propylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a white to off-white solid with a purity of ≥98.0%.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Solid appearance: White to off-white
  • Purity: ≥98.0%
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-60408 Supplier Medchemexpress LLC Supplier No. HYW007470100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a white to off-white solid with a purity of ≥98.0%.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Solid appearance: White to off-white
  • Purity: ≥98.0%
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.