Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propylboronic acid | 17745-45-8 | MFCD00069008 | 87.92 | 25 G
SDP

Catalog No. 5000449258
Encompass_Preferred

Propylboronic acid is a chemical reagent, typically appearing as a white to off-white solid with a purity of ≥98%. This compound is suitable for various research applications.

  • Purity of ≥98%
  • Appearance: White to off-white solid
  • Powder storage at -20°C for 3 years and 4°C for 2 years
  • In-solvent storage at -80°C for 6 months and -20°C for 1 month

Catalog No. 50-004-49258 Supplier Medchemexpress LLC Supplier No. 25GHYW007470
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propylboronic acid is a chemical reagent, typically appearing as a white to off-white solid with a purity of ≥98%. This compound is suitable for various research applications.

  • Purity of ≥98%
  • Appearance: White to off-white solid
  • Powder storage at -20°C for 3 years and 4°C for 2 years
  • In-solvent storage at -80°C for 6 months and -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.