Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propylcyclohexane

Catalog No. 111856100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-185-6100G 100 g
11-185-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-185-6100G Supplier Sigma Aldrich Supplier No. 111856100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propylcyclohexane is a component of the surrogate biokerosene model fuel
  • It has been used in the kinetic studies of kerosene jet a-1 oxidation
  • It has also been used to assay kerosene/rapeseed oil methyl ester (rme) mixture (80/20, mol/mol; biokerosene)

Specifications

CAS 1678-92-8
Density 0.793 g/mL (at 25°C (literature))
Boiling Point 155°C (lit.)
Molecular Formula C9H18
Refractive Index n20/D 1.436 (literature)
Linear Formula C6H11CH2CH2CH3
MDL Number MFCD00013770
Quantity 100 g
Molecular Weight (g/mol) 126.24
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.