Learn More
Sigma Aldrich Fine Chemicals Biosciences Propylene carbonate anhydr
Catalog No. 502898915
Encompass
Propylene carbonate is a cyclic carbonate that is commonly used as a solvent and as a reactive intermediate in organic synthesis. It is being considered as a potential electrochemical solvent due to its low vapor pressure high dielectric constant and high chemical stability.
Catalog No. 50-289-8915 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 310328100ML
Propylene carbonate is a cyclic carbonate that is commonly used as a solvent and as a reactive intermediate in organic synthesis. It is being considered as a potential electrochemical solvent due to its low vapor pressure high dielectric constant and high chemical stability.