Learn More
Sigma Aldrich Fine Chemicals Biosciences Propylene carbonate anhydr1L
Catalog No. 502899080
Encompass
Propylene carbonate is a cyclic carbonate that is commonly used as a solvent and as a reactive intermediate in organic synthesis. It is being considered as a potential electrochemical solvent due to its low vapor pressure high dielectric constant and high chemical stability.
Catalog No. 50-289-9080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3103281L
Propylene carbonate is a cyclic carbonate that is commonly used as a solvent and as a reactive intermediate in organic synthesis. It is being considered as a potential electrochemical solvent due to its low vapor pressure high dielectric constant and high chemical stability.