Learn More
Medchemexpress LLC Propylene glycol (1,2-propanediol) | 57-55-6 | MFCD00064272 | 76.09 g/mol | C3H8O2 | 1000mL
SDP
Catalog No. 5000269924
Encompass_Preferred
( )-1 2-propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol that is often used as an excipient in many active molecular preparations to increase the solubility and stability of the active molecule ( )-1 2-propanediol can affect the neurobehavior of zebrafish[1][2]
Catalog No. 50-002-69924 Supplier Medchemexpress LLC Supplier No. HYY09211000ML
( )-1 2-propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol that is often used as an excipient in many active molecular preparations to increase the solubility and stability of the active molecule ( )-1 2-propanediol can affect the neurobehavior of zebrafish[1][2]