Learn More
U.S. Pharmacopeia Propylene glycol, 57-55-6, MFCD00064272, 1 mL
Catalog No. NC9641737
Encompass
Linear Formula: CH3CH(OH)CH2OH, Molecular Weight: 76.09, BP: 187 °C, MP: −60 °C, Synonym: 1,2-Propanediol, Propylene glycol.
Catalog No. NC9641737 Supplier U.S. Pharmacopeia Supplier No. 1576708
Linear Formula: CH3CH(OH)CH2OH, Molecular Weight: 76.09, BP: 187 °C, MP: −60 °C, Synonym: 1,2-Propanediol, Propylene glycol.