Learn More
Medchemexpress LLC Propylene glycol | 57-55-6 | MFCD00064272 | 76.09 g/mol | C3H8O2 | 500mL
SDP
Catalog No. 5000268650
Encompass_Preferred
( )-1 2-propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol that is often used as an excipient in many active molecular preparations to increase the solubility and stability of the active molecule ( )-1 2-propanediol can affect the neurobehavior of zebrafish[1][2]
Catalog No. 50-002-68650 Supplier Medchemexpress LLC Supplier No. HYY0921500ML
( )-1 2-propanediol (1 2-(RS)-Propanediol) is an aliphatic alcohol that is often used as an excipient in many active molecular preparations to increase the solubility and stability of the active molecule ( )-1 2-propanediol can affect the neurobehavior of zebrafish[1][2]