Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol, FCC, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18605907 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-907 20 L Poly Bottle
18-605-904 500 mL Poly Bottle
18-605-908 6 x 500 mL Poly Bottle
18-605-905 4 L Poly Bottle
18-605-906 4 x 4 L Poly Bottle
18-605-909 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-907 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P145620LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1456, 57-55-6, C3H8O2

Spectrum™ Chemical Propylene Glycol, FCC is used as a humectant, solvent, and a preservative in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

CAS Max % 1
Boiling Point 188°C (370.4°F)
Ignition Residue 7e-005
Packaging Poly Bottle
Quantity 20 L
Percent Purity 99.5%
Grade FCC
Physical Form Liquid
Chemical Name or Material Propylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.