Learn More
Sigma Aldrich Fine Chemicals Biosciences Propylene glycol fCC fG
Catalog No. 502902369
Encompass
Propylene glycol is an aliphatic dihydric alcohol with a wide range of applications in the food industry such as an anticaking agent antioxidant humectant flavoring agent emulsifier stabilizer thickener etc.
Catalog No. 50-290-2369 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W29400410KGK
Propylene glycol is an aliphatic dihydric alcohol with a wide range of applications in the food industry such as an anticaking agent antioxidant humectant flavoring agent emulsifier stabilizer thickener etc.