Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propylene Glycol Monomethyl Ether Acetate

Catalog No. 48443118L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-443-118L 18 L
48-443-14L 4 L
48-443-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 48-443-118L Supplier Sigma Aldrich Supplier No. 48443118L
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Propylene glycol monomethyl ether acetate (PGMEA) is a photoresist solvent. Its degradation by microorganisms in different soil types has been investigated. An oral reference dose (RfD) value of PGMEA has been obtained from inhalation studies. The solubility of (5-alkylsulfonyloxyimino-5H-thiophen-2-ylidene)-2-methylphenyl-acetonitriles in PGMEA has been analyzed.Propylene glycol monomethyl ether acetate has been used in the development of fluidic channels to be incorporated in a poly(dimethylsiloxane) (PDMS)-based microfluidic device.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 108-65-6
Density 0.970 g/mL at 25°C
Boiling Point 145°C to 146°C; 145°C to 146°C
Molecular Formula C6H12O3
Refractive Index n20/D 1.402
Linear Formula CH3CO2CH(CH3)CH2OCH3
MDL Number MFCD00038500
Quantity 18 L
Synonym DOWANOL(R) PMA; 1,2-Propanediol monomethyl ether acetate; 1-Methoxy-2-propyl acetate; MPA; PGMEA; Propylene glycol methyl ether acetate
Molecular Weight (g/mol) 132.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.