Learn More
Electron Microscopy Sciences Propylene Glycol, Reagent A.C.S. 450 ML

Supplier: Electron Microscopy Sciences 20350
(1,2-Propanediol)
CAS #57-55-6 CH₃CHOHCH₂OH F.W. 76.10
Assay: >99.5%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.