Learn More
Description
Propylene Glycol USP is a clear, colorless, viscous liquid. It is practically odorless. Absorbs moisture when exposed to moist air.
Alternative name(s): 1,2-Propanediol, Monopropylene glycol, Mono propylene glycol, Propylene, Propane-1,2-diol, Propan-1,2-diol, methylethyl glycol
Specifications
Specifications
| Chemical Name or Material | Propylene Glycol USP |
| Packaging | Jug, 4L, White, HDPE, Square, 38-400, TE Pano Neck |
| CAS | 57-55-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 76.09 |
| Quantity | 4 L |
| Percent Purity | ≥99.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
