Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol, USP, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18605917 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
4 x 4 L
200 L
Packaging:
Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-917 200 L Poly Bottle
18-605-912 4 L Glass Bottle
18-605-913 4 x 4 L Glass Bottle
18-605-914 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-917 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR130200LTBL

missing translation for 'validMainframeMsg'

PR130, 57-55-6, C3H8O2

Spectrum™ Chemical Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

Acidity 0.20 mL
CAS Max % 1
Boiling Point 188°C (370.4°F)
Ignition Residue 3.5 mg/50g
Packaging Poly Bottle
Quantity 200 L
Percent Purity 99.5%
Grade USP
Physical Form Liquid
Chemical Name or Material Propylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.