Learn More
Polysciences, Inc. Propylene Oxide, EM Grade, 1 pint
SDP
Catalog No. NC0373394
Encompass_Preferred
Propylene Oxide, EM Grade, Solvent used in the last stage of dehydration of tissue for epoxy embedding. 75-56-9, 1 pint
Catalog No. NC0373394 Supplier Polysciences, Inc. Supplier No. 002361
Propylene Oxide, EM Grade, Solvent used in the last stage of dehydration of tissue for epoxy embedding. 75-56-9, 1 pint